Learn More
ACROBiosystems FcRn FCGRT B2M 2Vials
Catalog No. 502535268
Encompass_Preferred
FCGRT B2M heterodimer protein (FcRn complex) consist of two subunits p51 (equivalent to FCGRT) and p14 (equivalent to beta-2-microglobulin) and forms an MHC class I-like heterodimer. Fc fragment of IgG receptor transporter alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Catalog No. 50-253-5268 Supplier ACROBiosystems Supplier No. CHEKATP1322VIALS
FCGRT B2M heterodimer protein (FcRn complex) consist of two subunits p51 (equivalent to FCGRT) and p14 (equivalent to beta-2-microglobulin) and forms an MHC class I-like heterodimer. Fc fragment of IgG receptor transporter alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.