Learn More
ACROBiosystems HEK293/Human FcRn (FCGRT & B2M) Stable Cell Line, 2 vials (CHEK-ATP079-2VIALS)
Catalog No. 502357356
Encompass_Preferred
HEK293/Human FcRn (FCGRT & B2M) Stable Cell Line (CHEK-ATP079-2VIALS) is a cryopreserved HEK293 cell line engineered to stably express full-length human FCGRT (neonatal Fc receptor, FcRn) and beta-2-microglobulin (B2M). Surface expression of FcRn has been confirmed by flow cytometry, and the cells are intended for cell-based FcRn binding assays and bioassay development.
  • Engineered to stably express full-length human FCGRT and B2M.
  • Surface expression confirmed by flow cytometry.
  • Supplied as two cryopreserved vials with ≥5 x 10^6 cells per vial in 1 mL serum-free cryopreservation medium.
  • Stored in liquid nitrogen for long-term preservation.
  • Validated for cell-based FcRn (FCGRT & B2M) binding assays and cell-based bioassay development.
  • Mycoplasma testing negative and sterility testing negative.
  • Full traceable records and validated cell passage stability to support reproducible assays.
Catalog No. 50-235-7356 Supplier ACROBiosystems Supplier No. CHEKATP0792VIALS
HEK293/Human FcRn (FCGRT & B2M) Stable Cell Line (CHEK-ATP079-2VIALS) is a cryopreserved HEK293 cell line engineered to stably express full-length human FCGRT (neonatal Fc receptor, FcRn) and beta-2-microglobulin (B2M). Surface expression of FcRn has been confirmed by flow cytometry, and the cells are intended for cell-based FcRn binding assays and bioassay development.
  • Engineered to stably express full-length human FCGRT and B2M.
  • Surface expression confirmed by flow cytometry.
  • Supplied as two cryopreserved vials with ≥5 x 10^6 cells per vial in 1 mL serum-free cryopreservation medium.
  • Stored in liquid nitrogen for long-term preservation.
  • Validated for cell-based FcRn (FCGRT & B2M) binding assays and cell-based bioassay development.
  • Mycoplasma testing negative and sterility testing negative.
  • Full traceable records and validated cell passage stability to support reproducible assays.