Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque FcRn / FCGRT&B2M Heterodimer Protein, His Tag (BLI verified)
Catalog No. 501956689
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q8SPV9-1 (FCGRT) & Q8SPW0-1 (B2M)). In the region Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M), the AA sequence of Cynomolgus and Rhesus macaque FCGRT&B2M are homologus.;Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.1 kDa (FCGRT) and 11.6 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) contains no tag at the C-terminus. The reducing (R) protein migrates as 34 kDa (FCGRT) and 7 kDa (B2M) respectively due to glycosylation.;Captured Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified) (Cat. No. FCM-C52W9) on CM5 Chip via Anti-His antibody can bind Herceptin
Catalog No. 50-195-6689 Supplier ACROBiosystems Supplier No. FCMC52W91MG
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q8SPV9-1 (FCGRT) & Q8SPW0-1 (B2M)). In the region Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M), the AA sequence of Cynomolgus and Rhesus macaque FCGRT&B2M are homologus.;Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.1 kDa (FCGRT) and 11.6 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) contains no tag at the C-terminus. The reducing (R) protein migrates as 34 kDa (FCGRT) and 7 kDa (B2M) respectively due to glycosylation.;Captured Cynomolgus / Rhesus macaque FCGRT&B2M Heterodimer Protein, His Tag (BLI verified) (Cat. No. FCM-C52W9) on CM5 Chip via Anti-His antibody can bind Herceptin