Learn More
Proteintech Group Inc FcRn-Specific Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 161901AP150UL
Encompass_Preferred
The FcRn-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human FcRn-Specific. This antibody recognizes human antigen. The FcRn-Specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Catalog No. 50-225-6406