Learn More
ACROBiosystems Recombinant Protein;Biotinylated Feline FcRn / FCGRT & B2M Heterodimer Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;FCN,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems FCNF82W325UG
Encompass_Preferred
Recombinant Protein;Biotinylated Feline FcRn / FCGRT & B2M Heterodimer Protein, His Tag & Avi Tag (Avitag™);25UG;MABSol Biotinylated Feline FcRn / FCGRT & B2M Heterodimer Protein (FCN-F82W3) is expressed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) & Val 21 - Met 118 (B2M) (Accession # W5VRL5-1 (FCGRT) & Q5MGS7-1 (B2M)).;The subunit FCGRT carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 34 kDa (FCGRT), 11.5 kDa (B2M) . The protein migrates as 36-40 kDa (FCGRT), 18-20 kDa (B2M) respectively on a SDS-PAGE gel under reducing (R) condition due to glycosylation.
Catalog No. 50-165-8141