Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque FcRn / FCGRT & B2M Heterodimer Protein (SPR verified);HEK293;1MG;FCM,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems FCMC52841MG
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque FcRn / FCGRT & B2M Heterodimer Protein (SPR verified);1MG;Cynomolgus / Rhesus macaque FcRn Heterodimer Protein (SPR verified), is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q8SPV9-1 (FCGRT) & Q8SPW0-1 (B2M)).;Cynomolgus / Rhesus macaque FcRn Heterodimer Protein (SPR verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 32.3 kDa (FCGRT) and 13.1 kDa (B2M). Subunit FCGRT is fused with a polyhistidine tag at the C-terminus and subunit Beta-2 microglobulin (B2M) is fused with Strep II-tag at the C-terminus. The reducing (R) protein migrates as 36 kDa (FCGRT) and 14 kDa (B2M) respectively due to glycosylation. Cynomolgus / Rhesus macaque FcRn Heterodimer Protein (SPR verified) sequences are identical.;Immobilized Anti-Human CTLA4 Mab at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque FcRn Heterodimer Protein (SPR verifi
Catalog No. 50-165-9135