Learn More
ACROBiosystems Recombinant Protein;Biotinylated Rat FcRn/FCGRT & B2M, Avi Tag (Avitag™);HEK293;200UG;FCM,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Catalog No. 501659136
Encompass_Preferred
Recombinant Protein;Biotinylated Rat FcRn/FCGRT & B2M, Avi Tag (Avitag™);200UG;MABSol Biotinylated Rat FcRn / FCGRT & B2M Heterodimer Protein (FCM-R82W7) is expressed from human HEK293 cells. It contains AA Ala 23 - Ser 298 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # NP_203502 (FcRN) & NP_036644 (B2M)).;The subunit FCGRT carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The subunit Beta-2 microglobulin (B2M) carries a Strep II tag at the C-terminus. The protein has a calculated MW of 34.7 kDa (FCGRT), 13.1 kDa (B2M) . The protein migrates as 45-50kDa (FCGRT), 14 kDa (B2M) respectively on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Yervoy Ipilimumab at 5 g/mL (100 L/well) can bind Biotinylated Rat FcRn / FCGRT & B2M (Cat. No. FCM-R82W7) with a linear range of 0.02-0.15 g/mL (QC tested).
Catalog No. 50-165-9136 Supplier ACROBiosystems Supplier No. FCMR82W7200UG
Recombinant Protein;Biotinylated Rat FcRn/FCGRT & B2M, Avi Tag (Avitag™);200UG;MABSol Biotinylated Rat FcRn / FCGRT & B2M Heterodimer Protein (FCM-R82W7) is expressed from human HEK293 cells. It contains AA Ala 23 - Ser 298 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # NP_203502 (FcRN) & NP_036644 (B2M)).;The subunit FCGRT carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The subunit Beta-2 microglobulin (B2M) carries a Strep II tag at the C-terminus. The protein has a calculated MW of 34.7 kDa (FCGRT), 13.1 kDa (B2M) . The protein migrates as 45-50kDa (FCGRT), 14 kDa (B2M) respectively on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Yervoy Ipilimumab at 5 g/mL (100 L/well) can bind Biotinylated Rat FcRn / FCGRT & B2M (Cat. No. FCM-R82W7) with a linear range of 0.02-0.15 g/mL (QC tested).