Learn More
ORS Nasco FCSHLD REP VSR BIONIC CL
Catalog No. 502551566
Encompass_Preferred
Honeywell part# B074GI/L. Honeywell North 7 mil Butyl Chemical Resistant Gloves Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Size: Large Style: North Butyl B074GI Thickness: 7 mil 0.17 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR MIL-DTL-43976D EN 374 EN 420 Grip Design: Smooth
Catalog No. 50-255-1566 Supplier ORS Nasco Supplier No. 763S8555
Honeywell part# B074GI/L. Honeywell North 7 mil Butyl Chemical Resistant Gloves Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Size: Large Style: North Butyl B074GI Thickness: 7 mil 0.17 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR MIL-DTL-43976D EN 374 EN 420 Grip Design: Smooth