Learn More
Sorbent Technologies Inc Flash Cartridge ver. 2, Screw Top, Luer Lock end fittings, Includes top and bottom frit, o-ring, dispersing insert, PP, 220g, 376ml, 6/pk
SDP
Supplier:  Sorbent Technologies Inc FCSTLLV22206
Encompass_Preferred
These empty flash cartridges come ready for you to easily pack and repack them with any adsorbent. This solvent resistant polypropylene cartridge has a liquid volume of 303ml. Cartridges are rated at 150 psi (10.3 bar). The cartridge has a luer lock connection at the top and bottom. Pack includes cartridges, top and bottom frits, o-rings, and dispersing inserts. For use with Sorbent Technologies' Silica Gels, Aluminas, and other adsorbents.
Catalog No. NC2962626