Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FD-1080 free acid | 1151888-25-3 | 96.3% | C40H39ClN2O6S2 | 10MG
SDP

Catalog No. 5000259035
Encompass_Preferred

FD-1080 free acid is a near-infrared II (NIR-II) small-molecule fluorophore with excitation at 1064 nm and emission at 1080 nm, optimized for deep-tissue in vivo imaging. Supplied as a solid free acid with high purity and DMSO solubility, it provides photostability and compatibility with NIR-II imaging systems.

  • Excitation at 1064 nm and emission at 1080 nm for NIR-II imaging.
  • High purity (96.3%) for reliable experimental results.
  • Soluble in DMSO (20 mg/mL) with ultrasonic assistance.
  • Solid free acid form suitable for storage and handling.
  • Photostable under continuous laser irradiation.
  • Store at -80°C, protect from light, and keep under inert atmosphere.

Catalog No. 50-002-59035 Supplier Medchemexpress LLC Supplier No. HY133852A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

FD-1080 free acid is a near-infrared II (NIR-II) small-molecule fluorophore with excitation at 1064 nm and emission at 1080 nm, optimized for deep-tissue in vivo imaging. Supplied as a solid free acid with high purity and DMSO solubility, it provides photostability and compatibility with NIR-II imaging systems.

  • Excitation at 1064 nm and emission at 1080 nm for NIR-II imaging.
  • High purity (96.3%) for reliable experimental results.
  • Soluble in DMSO (20 mg/mL) with ultrasonic assistance.
  • Solid free acid form suitable for storage and handling.
  • Photostable under continuous laser irradiation.
  • Store at -80°C, protect from light, and keep under inert atmosphere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.