Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Yellow No. 5, Spectrum™ Chemical
SDP

Catalog No. 18602640 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-640 25 kg Amber Glass Bottle
18-602-636 100 g Amber Glass Bottle
18-602-637 500 g Amber Glass Bottle
18-602-638 2.5 kg Amber Glass Bottle
18-602-639 20 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-640 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. FD15025KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FD150, 1934-21-0, C16H9N4O9S2Na3

Spectrum™ Chemical FD and C Yellow No. 5

Chemical Identifiers

CAS 1934-21-0
Molecular Formula C16H9N4Na3O9S2
Molecular Weight (g/mol) 534.36
InChI Key UJMBCXLDXJUMFB-UHFFFAOYNA-K
IUPAC Name trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate
SMILES [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Color Orange
pH 6 to 8
Quantity 25 kg
Percent Purity 87%
Grade Reagent
Chemical Name or Material FD and C Yellow No. 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.