Learn More
LAB ALLEY Lab Alley Food Grade Ethanol 200 Proof Kosher Halal
SDP
Catalog No. NC2373917
Encompass
Pure sometimes called absolute ethanol is 100% ethanol. Absolute food grade ethanol is preferred for many common techniques in food research and development including food grade extraction and fraction isolation or food grade synthesis. For food grade synthesis the presence of water could result in undesired byproducts. For example many volatile organic compounds are synthesized in ethanol and can be added to food to manipulate flavor and aroma. The use of food grade solvents in all of these applications ensures the safety and purity of food and food standards for consumers. Buy Food Grade Ethanol 200 Proof 100% Pure Alcohol In Bulk Sizes Non-Denatured Ethyl Alcohol Products For Sale Packaged In Bottles Drums Totes Pallets.
Catalog No. NC2373917 Supplier LAB ALLEY Supplier No. EAF2001LCS
Pure sometimes called absolute ethanol is 100% ethanol. Absolute food grade ethanol is preferred for many common techniques in food research and development including food grade extraction and fraction isolation or food grade synthesis. For food grade synthesis the presence of water could result in undesired byproducts. For example many volatile organic compounds are synthesized in ethanol and can be added to food to manipulate flavor and aroma. The use of food grade solvents in all of these applications ensures the safety and purity of food and food standards for consumers. Buy Food Grade Ethanol 200 Proof 100% Pure Alcohol In Bulk Sizes Non-Denatured Ethyl Alcohol Products For Sale Packaged In Bottles Drums Totes Pallets.