Learn More
Medchemexpress LLC FD-IN-1 | 1646682-14-5 | 98.6% | 100 MG
SDP
Catalog No. 5000338273
Encompass_Preferred
FD-IN-1 (Compound 12) is an orally bioavailable and selective factor D (FD) inhibitor with an IC50 of 12 nM. Complement FD, a highly specific S1 serine protease, plays a central role in the alternative complement pathway of the innate immune system. FD-IN-1 also inhibits factor XIa (FXIa) and Tryptase β2 with IC50s of 7.7 μM and 6.5 μM, respectively. It appears as a white to off-white solid.
  • Selective factor D inhibition
  • Orally bioavailable
  • Systemic and local suppression of AP activation
  • Research use
  • Potential for autoimmune and inflammatory diseases
Catalog No. 50-003-38273 Supplier Medchemexpress LLC Supplier No. HY128570100MG
FD-IN-1 (Compound 12) is an orally bioavailable and selective factor D (FD) inhibitor with an IC50 of 12 nM. Complement FD, a highly specific S1 serine protease, plays a central role in the alternative complement pathway of the innate immune system. FD-IN-1 also inhibits factor XIa (FXIa) and Tryptase β2 with IC50s of 7.7 μM and 6.5 μM, respectively. It appears as a white to off-white solid.
  • Selective factor D inhibition
  • Orally bioavailable
  • Systemic and local suppression of AP activation
  • Research use
  • Potential for autoimmune and inflammatory diseases