Learn More
Fd Neurotechnologies Inc FD RAPID GOLGISTAIN KIT SMALL
SDP
Supplier:  Fd Neurotechnologies Inc PK401A
Encompass
Using the Golgi technique, subtle morphological alterations in neuronal dendrites and dendritic spines have been discovered in the brains of animals treated with drugs as well as in the postmortem brains of patients with neurological diseases3, 4. However, the unreliability and the time-consuming process of Golgi staining have been major obstacles to the widespread application of this technique. FD Rapid GolgiStain™ Kit is designed based on the principle of the methods described by Ramón- Moliner2, Glaser and Van der Loos5. This kit has not only dramatically improved and simplified the Golgi-Cox technique but has also proven to be extremely reliable and sensitive for demonstrating morphological details of neurons and glia, especially dendritic spines. The FD Rapid GolgiStain™ Kit has been tested extensively and widely used on the brains from several species of animals as well as on the specimens of postmortem human brains
Catalog No. NC0292960