Learn More
Fd Neurotechnologies Inc FD Section Storage Solution™ 250mL
SDP
Catalog No. NC1574336
Encompass
The FD Section Storage Solution™ is a specifically formulated buffer solution that stays liquid at -20°C. It is ideal for the prolonged storage of free-floating fixed tissue sections. This solution has been proven to preserve the best morphology and antigenicity for cryostat and vibratome sections. Sections kept in this solution may be stored at -20°C for at least 5 years before processing for immunohistochemistry or routine histology.
Catalog No. NC1574336 Supplier Fd Neurotechnologies Inc Supplier No. PC101
The FD Section Storage Solution™ is a specifically formulated buffer solution that stays liquid at -20°C. It is ideal for the prolonged storage of free-floating fixed tissue sections. This solution has been proven to preserve the best morphology and antigenicity for cryostat and vibratome sections. Sections kept in this solution may be stored at -20°C for at least 5 years before processing for immunohistochemistry or routine histology.