Learn More
Fd Neurotechnologies Inc FD Tissue Cryoprotection Solution™, 250mL
SDP
Catalog No. NC3763024
Encompass
FD Tissue Cryoprotection Solution™ is a specifically formulated buffer solution that prevents tissue from ice crystal damage during freezing* and preserves the best possible morphology and antigenicity of cells. It has been proven to greatly facilitate both sectioning and various histological stains. In addition, tissue treated with this solution can be easily cut as thick as 300 microns on either a cryostat or a sliding microtome without shattering. The solution may be used for treatment of tissues fixed with any fixative.
Catalog No. NC3763024 Supplier Fd Neurotechnologies Inc Supplier No. PC102
FD Tissue Cryoprotection Solution™ is a specifically formulated buffer solution that prevents tissue from ice crystal damage during freezing* and preserves the best possible morphology and antigenicity of cells. It has been proven to greatly facilitate both sectioning and various histological stains. In addition, tissue treated with this solution can be easily cut as thick as 300 microns on either a cryostat or a sliding microtome without shattering. The solution may be used for treatment of tissues fixed with any fixative.