Learn More
Fd Neurotechnologies Inc FD TISSUE STORAGE SOLU 250ML
SDP
Catalog No. NC2013560
Encompass
FD Tissue Storage Solution is specifically designed for the prolonged storage of tissues, especially brains and spinal cords that have been fixed with formaldehyde.
Catalog No. NC2013560 Supplier Fd Neurotechnologies Inc Supplier No. PC103
Add to Cart
Add to Cart
FD Tissue Storage Solution is specifically designed for the prolonged storage of tissues, especially brains and spinal cords that have been fixed with formaldehyde.