Learn More
Julabo USA Inc Each, FD200 flow-through cooler 220W
SDP
Catalog No. 507534002
Encompass_Preferred
Chillers, Julabo, FD200 Flow-Through Cooler +10...+30 °C. FD200 flow-through cooler is the perfect complement to open bath circulators, immersion circulators and bridge-mounted circulators extending their temperature range to below ambient temperature. It offers a small footprint, low energy requirement, and represents an economic solution preventing tap water usage for cooling. Instruments are designed for integration into an existing cooling loop (e.g. consisting of open bath circulator with pump). The FD200 flow through cooler is used for cooling a loop circuit of a closed system, the unit must be connected in the return line of a heating circulator. The compact, easy to use system is suitable for a variety of cooling applications. Simple operation with a range of +10…+30 °C with 220 W of cooling power at 20 °C.
Catalog No. 50-753-4002 Supplier Julabo USA Inc Supplier No. 9655825.2
Chillers, Julabo, FD200 Flow-Through Cooler +10...+30 °C. FD200 flow-through cooler is the perfect complement to open bath circulators, immersion circulators and bridge-mounted circulators extending their temperature range to below ambient temperature. It offers a small footprint, low energy requirement, and represents an economic solution preventing tap water usage for cooling. Instruments are designed for integration into an existing cooling loop (e.g. consisting of open bath circulator with pump). The FD200 flow through cooler is used for cooling a loop circuit of a closed system, the unit must be connected in the return line of a heating circulator. The compact, easy to use system is suitable for a variety of cooling applications. Simple operation with a range of +10…+30 °C with 220 W of cooling power at 20 °C.