Learn More
Enzo Life Sciences SCREEN-WELL FDA approved drug library V2 (100µl/well)
SDP
Catalog No. 502010218
Encompass_Preferred
Drug repositioning or repurposing can be an important part of any drug discovery program and has led to several blockbuster drugs (e.g. Viagra and Rogaine). High-content screens, new biomarkers and noninvasive imaging techniques have created new opportunities for pursuing novel indications for approved compounds. Being FDA approved and in use in the clinics, all of the compounds in SCREEN-WELL FDA v. 2.0 Approved Drug Library have known and well-characterized bioactivity, safety and bioavailability - properties which could dramatically accelerate drug development and optimization. The library also avoids irrelevant compounds found in similar libraries, such as herbicides, insecticides, sunscreen agents, and cytotoxic agents. This set provides one of the richest sources of bioactives and offers the greatest degree of drug-likeness available. Hits from this set will provide a significant head start in any drug optimization program. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0218 Supplier Enzo Life Sciences Supplier No. BML28430100
Add to Cart
Add to Cart
Request a Quote
Drug repositioning or repurposing can be an important part of any drug discovery program and has led to several blockbuster drugs (e.g. Viagra and Rogaine). High-content screens, new biomarkers and noninvasive imaging techniques have created new opportunities for pursuing novel indications for approved compounds. Being FDA approved and in use in the clinics, all of the compounds in SCREEN-WELL FDA v. 2.0 Approved Drug Library have known and well-characterized bioactivity, safety and bioavailability - properties which could dramatically accelerate drug development and optimization. The library also avoids irrelevant compounds found in similar libraries, such as herbicides, insecticides, sunscreen agents, and cytotoxic agents. This set provides one of the richest sources of bioactives and offers the greatest degree of drug-likeness available. Hits from this set will provide a significant head start in any drug optimization program. Long Term Storage: -80°C. Applications: HTS .