Learn More
MSC Vikan Ultra Hygiene Table Squeegee, 10 in Blade, TPE Rubber, Purple, FDA Compliant
Supplier:  MSC 47167531
Encompass
This ultra-hygienic single-bladed squeegee is designed for removing excess water and food debris from food-preparation surfaces, minimizing areas for soils to collect. Its soft and flexible TPE rubber blade is fully molded with smooth, rounded surfaces for better hygiene and easier cleaning. Made from FDA-compliant materials, this squeegee is autoclavable up to 250°F (121°C) and dishwasher safe, making it ideal for environments requiring the highest level of hygiene and strict cross-contamination control.
  • Fully molded single-blade design
  • Smooth, rounded surfaces for improved hygiene
  • Easy to clean
  • Soft, flexible TPE rubber blade
  • Elongated body reduces blade wear
  • Removes excess water and food debris
  • Made from FDA-compliant materials
  • Autoclavable up to 250°F (121°C)
  • Dishwasher safe
  • Color-coding compatible for cross-contamination prevention
Catalog No. 50-283-5516