Learn More
Amsbio LLC FDA normal organ tissue array
Catalog No. 502564502
Encompass
FDA normal organ tissue array of cynomolgus monkey 20 cases/32 types/99 cores (core size 1.0mm) replacing CyFDA1g
Catalog No. 50-256-4502 Supplier Amsbio LLC Supplier No. CYFDA1H
FDA normal organ tissue array of cynomolgus monkey 20 cases/32 types/99 cores (core size 1.0mm) replacing CyFDA1g