Learn More
Cayman Chemical FDAApproved Drugs Screeng 25uL
SDP
Catalog No. 502995304
Encompass_Preferred
A potent inhibitor of E-selectin and ICAM-1 expression in HUVECs stimulated with TNF-a (IC50s 20 and 25 nM respectively)
Catalog No. 50-299-5304 Supplier Cayman Chemical Supplier No. 2353825 UL
A potent inhibitor of E-selectin and ICAM-1 expression in HUVECs stimulated with TNF-a (IC50s 20 and 25 nM respectively)