Learn More
Medchemexpress LLC HY-112721 10mg Medchemexpress, FDI-6 CAS:313380-27-7 Purity:>98%
SDP
Catalog No. 501969156
Encompass_Preferred
Medchemexpress, HY-112721 10mg FDI-6 CAS:313380-27-7 FDI-6 is an inhibitor of FOXM1. FDI-6 binds directly to FOXM1 protein, to displace FOXM1 from genomic targets in MCF-7 breast cancer cells, and induce concomitant transcriptional down-regulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9156 Supplier Medchemexpress LLC Supplier No. HY11272110MG
Medchemexpress, HY-112721 10mg FDI-6 CAS:313380-27-7 FDI-6 is an inhibitor of FOXM1. FDI-6 binds directly to FOXM1 protein, to displace FOXM1 from genomic targets in MCF-7 breast cancer cells, and induce concomitant transcriptional down-regulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.