Learn More
Selleck Chemical LLC FDI-6-5mg
SDP
Catalog No. 502389931
Encompass_Preferred
FDI-6 (NCGC00099374) is an inhibitor of the Forkhead box protein M1 transcription factor (FOXM1) with IC50 of 22.5 μM.
Catalog No. 50-238-9931 Supplier Selleck Chemical LLC Supplier No. S96895MG
FDI-6 (NCGC00099374) is an inhibitor of the Forkhead box protein M1 transcription factor (FOXM1) with IC50 of 22.5 μM.