Learn More
Waters Corp Preventative Maintenance Kit, Fluid Delivery Module, SFC
Catalog No. 502391964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preventative maintenance kit is designed for the Waters Fluid Delivery Module (FDM) to ensure optimal performance and longevity. It is compatible with Investigator SFC System, Prep 15 SFC Purification System, SFC Prep 15, and Fluid Delivery Module 10 systems.
  • Contains multiple seals and backup rings for the fluid delivery module.
  • Includes various check valves for critical fluid handling.
  • Provides replacement filters and filter elements for the fluid delivery module.
  • Supplies a piston seal kit for maintenance.
  • Features a seal for the prime/purge valve.
  • Comes with an inline replacement check valve cartridge.
Catalog No. 50-239-1964 Supplier Waters Corp Supplier No. 700007690
Add to Cart
Add to Cart
Request a Quote
This preventative maintenance kit is designed for the Waters Fluid Delivery Module (FDM) to ensure optimal performance and longevity. It is compatible with Investigator SFC System, Prep 15 SFC Purification System, SFC Prep 15, and Fluid Delivery Module 10 systems.
  • Contains multiple seals and backup rings for the fluid delivery module.
  • Includes various check valves for critical fluid handling.
  • Provides replacement filters and filter elements for the fluid delivery module.
  • Supplies a piston seal kit for maintenance.
  • Features a seal for the prime/purge valve.
  • Comes with an inline replacement check valve cartridge.