Learn More
Shelby™ FDP™ Brushed Pigskin Leather Gloves - Knit Wrist
Catalog No. 19821480 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-480 Medium
19-100-374 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-821-480 Supplier Shelby™ Supplier No. 5225M
This item is not returnable. View return policy
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
  • Durable outer shells are made of either soft, koala-tanned cowhide leather or abrasion-resistant brushed pigskin
  • Gloves feature Gore RT7100 glove barrier fabric laminated to SEF modacrylic fleece
  • Lining is sewn to shell at fingertips and wrist to eliminate pullouts
  • Seams are sealed with DuPont™ silicone sealant
  • Gunn cut with wing thumb construction
  • Welted thumb and fourchette seams
  • Glove body extends 2 in. (5 cm) above wrist crease with shirred elastic at wrist
  • 0.25 in. (6.3 mm) hanger loop for easy storage
Specifications
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Tan
Material Pigskin Leather
Product Type Structural Firefighting Gloves
Size Medium
Glove Cuff Style Knit
Liner Material Thermal