Learn More
Aviva Systems Biology Corp FDPS Antibody - C-terminal region (ARP60781_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60781_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FDPS. It was validated on Western Blot. . Alternate names: FPPS, FPSApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-161