Learn More
Aviva Systems Biology Corp FDXR Antibody - N-terminal region (ARP54706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FDXR. It was validated on Western Blot. . Alternate names: ADXRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-957