Learn More
Strem, An Ascensus Company CAS# 14017-39-1. 250g. Iron(II) sulfamate, (38-42%) aqueous solution. MFCD00055536
SDP
Catalog No. 502045419
Encompass
CAS# 14017-39-1. 250g. Iron(II) sulfamate, (38-42%) aqueous solution. MFCD00055536. Molecular Weight: 248.02. Molecular Formula: Fe(NH2SO3)2. Color/form: green liq. Strem# 93-2638. http://www.strem.com/catalog/v/93-2638/
Catalog No. 50-204-5419 Supplier Strem, An Ascensus Company Supplier No. 932638250G
CAS# 14017-39-1. 250g. Iron(II) sulfamate, (38-42%) aqueous solution. MFCD00055536. Molecular Weight: 248.02. Molecular Formula: Fe(NH2SO3)2. Color/form: green liq. Strem# 93-2638. http://www.strem.com/catalog/v/93-2638/