Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fe/Ppm Pd For Nitro Group Reductions

Catalog No. 809330500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-933-0500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 80-933-0500MG Supplier Sigma Aldrich Supplier No. 809330500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

When these nanoparticles (NPs) are used along with PEG-containing designer surfactants such as TPGS-750-M, a facile reduction of nitro compounds can be effected in water at room temperature. The transformation is highly chemoselective and tolerates a large variety of functional groups.Watch Professor Lipshutz talk about Fe/ppm Pd in this webinar:From milligrams to kilograms: synthetic chemistry following nature's lead

Specifications

CAS 1603164-23-3
Linear Formula C16H22F6N3PRu
Quantity 500 mg
Synonym 25,26,27,28-Tetrahydroxycalix[4]arene-5,11,17,23-tetrasulfonic acid sodium salt; Calix[4]arene-4-sulfonic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.