Learn More
Nanocomposix NanoXact Magnetite Nanoparticles - PVP - 20 nm, 20 mg/mL, in aq. 2 mM sodium citrate sol., 5 mL
SDP
Supplier:  Nanocomposix MGPN205M
Encompass_Preferred
nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.20 nm magnetite (iron oxide) nanoparticles combine superparamagnetic properties at ambient temperatures with a high surface area to volume ratio. These nanoparticles are useful in a broad range of applications from nanomedicine, imaging contrast agents, drug delivery, hyperthermia therapy, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. 50-195-9462