Learn More
Andwin Scientific FEATHER DISP SCALPEL NO15 20PK
SDP
Catalog No. NC9967540
Encompass_Preferred
The Feather Disposable Scalpels offer the sharpest edge available, making it ideal for field use. Each scalpel is individually wrapped in aluminum wrapper to ensure its cleanliness. This product is sterile and is ready to use. It is also disposable in order to eliminate the chance of contamination. This product is in style #15.
Catalog No. NC9967540 Supplier Andwin Scientific Supplier No. EF7281C
The Feather Disposable Scalpels offer the sharpest edge available, making it ideal for field use. Each scalpel is individually wrapped in aluminum wrapper to ensure its cleanliness. This product is sterile and is ready to use. It is also disposable in order to eliminate the chance of contamination. This product is in style #15.