Learn More
Electron Microscopy Sciences Feather Double Edge Razor Blades
Supplier: Electron Microscopy Sciences 7200210
Made from carbon steel. Feather blades are well known to have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker and Holder. Blades are available in packages of 10 blades per box or 500 blades per box. Measurements: Width: 21.95 mm (0.865") x Length is 42.96 mm (1.69"). Thickness: 0.10 mm (0.004")