Learn More
Electron Microscopy Sciences Feather Double Edge Razor Blades
SDP
Catalog No. 50949135
Encompass_Preferred
Made from carbon steel. Feather blades are well known to have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker and Holder. Blades are available in packages of 10 blades per box or 500 blades per box. Measurements: Width: 21.95 mm (0.865") x Length is 42.96 mm (1.69"). Thickness: 0.10 mm (0.004")
Catalog No. 50-949-135 Supplier Electron Microscopy Sciences Supplier No. 7200201
Add to Cart
Add to Cart
Made from carbon steel. Feather blades are well known to have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker and Holder. Blades are available in packages of 10 blades per box or 500 blades per box. Measurements: Width: 21.95 mm (0.865") x Length is 42.96 mm (1.69"). Thickness: 0.10 mm (0.004")