Learn More
Electron Microscopy Sciences Feather™ Stainless Steel Micro Scalpel 45°, Plastic Handle
SDP
Catalog No. 50366044
Encompass_Preferred
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Plastic handle.
Angle: 45°
Catalog No. 50-366-044 Supplier Electron Microscopy Sciences Supplier No. 7204645
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Plastic handle.
Angle: 45°