Learn More
Electron Microscopy Sciences Feather Safeshield™ Blade #11
SDP
Catalog No. 50364922
Encompass_Preferred
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures. They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Catalog No. 50-364-922 Supplier Electron Microscopy Sciences Supplier No. 7204311
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures. They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.