Learn More
Electron Microscopy Sciences Scalpel Handle #3
SDP
Catalog No. 50949167
Encompass_Preferred
Scalpel handles for use with different sized blades. 100mm long, flat with corrugated grip, for use with scalpel blades #10, #11, #12 and #15.
Catalog No. 50-949-167 Supplier Electron Microscopy Sciences Supplier No. 7204003
Scalpel handles for use with different sized blades. 100mm long, flat with corrugated grip, for use with scalpel blades #10, #11, #12 and #15.