Learn More
Electron Microscopy Sciences Scalpel Handle #4
SDP
Catalog No. 50949168
Encompass_Preferred
Scalpel handles for use with different sized blades. 105mm long, flat with corrugated grip, for use with scalpel blades #20, #21, #22, #23, and #24.
Catalog No. 50-949-168 Supplier Electron Microscopy Sciences Supplier No. 7204004
Scalpel handles for use with different sized blades. 105mm long, flat with corrugated grip, for use with scalpel blades #20, #21, #22, #23, and #24.