Learn More
Electron Microscopy Sciences Scalpel Handle #7
SDP
Catalog No. 50949186
Encompass_Preferred
Scalpel handles for use with different sized blades. 140mm long, slender, contoured with corrugations, for use with scalpel blades #10, #11, #12, and #15.
Catalog No. 50-949-186 Supplier Electron Microscopy Sciences Supplier No. 7204007
Scalpel handles for use with different sized blades. 140mm long, slender, contoured with corrugations, for use with scalpel blades #10, #11, #12, and #15.