Learn More
Graham-Field™ Feather™ Sterile Surgical Blades
SDP
Catalog No. 19027496
Change view
Click to view available options
Size:
# 12
Catalog No. Size
19-027-496 # 12
1 options
This item is not returnable. View return policy
Catalog No. 19-027-496 Supplier Graham-Field™ Supplier No. 2976#12
Add to Cart
Add to Cart
This item is not returnable. View return policy
Superior cutting quality, strength and corrosion resistant characterize these stainless steel, disposable Feather™ Sterile Surgical Blades. Individually wrapped and sealed to maintain sterility (gamma irradiation) and insure long shelf-life.
  • Heat-treated stainless steel blades provide superior cutting quality, strength, and corrosion resistance
  • Gamma-irradiated sterile blades are single use
  • Color-coded packaging assists with blade identification
  • All blades are individually packaged 100 per dispensing box
Specifications
Blade Material Stainless Steel
Product Type Surgical Blade
Size # 12
Disposable Single-use
Sterility Sterile
Certifications/Compliance ISO 14001 Compliant, ISO 9001 Certified, ISO 13485 Certified
Color Orange
Quantity 100 Pk.
Packaging Type Individually Wrapped in a Dispensing Box