Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences MicroPoint™ Featherlite MPF-5, Smooth/Smooth, 140 mm
SDP

Catalog No. 5027232
Encompass_Preferred

MicroPoint scissors can be used for suture or tissue and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors cut by shearing and have three times the shearing angle to reduce "tissue crushing. They last far longer than ordinary scissors.

O.R. Castro-Viejo quality at a floor-grade price.
They offer consistent cutting at the tip.
The blades cannot work loose and the contact between the blades remains constant.
Adjustable shearing contact.
All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening.
Surface: Smooth
OAL: Needlenose Round Tip Straight
Blade: 15 mm
Blade Size: 140 mm

Catalog No. 50-272-32 Supplier Electron Microscopy Sciences Supplier No. 7791031
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroPoint scissors can be used for suture or tissue and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors cut by shearing and have three times the shearing angle to reduce "tissue crushing. They last far longer than ordinary scissors.

O.R. Castro-Viejo quality at a floor-grade price.
They offer consistent cutting at the tip.
The blades cannot work loose and the contact between the blades remains constant.
Adjustable shearing contact.
All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening.
Surface: Smooth
OAL: Needlenose Round Tip Straight
Blade: 15 mm
Blade Size: 140 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.