Learn More
Perkin Elmer US LLC Elegra Argon Humidifier for NexION 1000/2000
Supplier:  Perkin Elmer US LLC N8150493
Encompass_Preferred
Features and benefits include: An easy-to-use bypass switch allows you to take the Elegra off-line without disconnecting argon lines. Highly efficient membrane humidification technology. Improved signal stability for samples with high TDS. Simple to use and maintain. Facilitates long, uninterrupted run times. Inert metal-free construction eliminates possibility of contamination. Rugged and durable polymer casing. Maximum and minimum fill marks ensure that you are always operating under optimum conditions.
Catalog No. 50-201-6489