Learn More
Medchemexpress LLC Febantel | 58306-30-2 | 99.9% | 446.48 | 5 G
SDP
Catalog No. 5000387334
Encompass_Preferred
Febantel is an oral dewormer that treats gastrointestinal nematodes in sheep, as well as roundworm and tapeworm infections in poultry and livestock. It also effectively inhibits the embryonic development of mouse hairworms and whipworm eggs.
  • Treats gastrointestinal nematodes in sheep.
  • Treats roundworm and tapeworm infections in poultry and livestock.
  • Inhibits embryonic development of mouse hairworms and whipworm eggs.
Catalog No. 50-003-87334 Supplier Medchemexpress LLC Supplier No. HY175975G
Febantel is an oral dewormer that treats gastrointestinal nematodes in sheep, as well as roundworm and tapeworm infections in poultry and livestock. It also effectively inhibits the embryonic development of mouse hairworms and whipworm eggs.
  • Treats gastrointestinal nematodes in sheep.
  • Treats roundworm and tapeworm infections in poultry and livestock.
  • Inhibits embryonic development of mouse hairworms and whipworm eggs.