Learn More
Lumos Diagnostics FebriDx™ Bacterial / Non-Bacterial Point-of-Care Assay
Catalog No. 21-402-724
Change view
Click to view available options
Test Kit
Catalog No.
21-402-724 Test Kit
1 options
This item is not returnable. View return policy
Catalog No. 21-402-724 Supplier Lumos Diagnostics Supplier No. CP0014
This item is not returnable. View return policy
FebriDx is a rapid point-of-care test that uses a fingerstick blood sample to aid in the differentiation between bacterial infection and non-bacterial etiology.
  • FebriDx is a rapid point-of-care test that uses a fingerstick blood sample to aid in the differentiation between bacterial infection and non-bacterial etiology.
  • FebriDx is intended to be used in urgent and emergency care settings.Knowing whether a patient has a bacterial infection has a direct impact on reducing unnecessary antibiotic prescriptions, limiting the spread of antibiotic-resistant bacteria, and helping providers know when to initiate treatment.
  • Results after 10 minutes increases confidence in whether or not to prescribe an antibiotic.
  • Highly sensitive/specific dual biomarker technology provides reliable differentiation of bacterial and non-bacterial infections.
  • All-in-one, instrument-free test device means no expensive equipment and a fully portable solution. Increase confidence in decision making.
  • Confidently rule out a bacterial infection with 99% negative predictive value (NPV).
  • Ensure bacterial infections are not overlooked.
  • Increased confidence in patient diagnosis.
  • Improve patient satisfaction.
  • Support patient understanding of treatment plan with a tangible test result.
  • Enable improved patient management.
  • Actionable results after 10 minutes for use during a patient visit.
  • Support antimicrobial stewardship.
  • Reduce unnecessary antibiotic use.
Specifications
CE Marker No
Clia Complexity Moderate
Detectable Analytes Influenza A and Influenza B Nucleoprotein Antigens
Type Point-of-Care Assay
Disposable Single-use
Form Ready-to-Use (RTU)