Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lumos Diagnostics FebriDx™ External Controls

Catalog No. 21200227
Change view
Click to view available options
External Controls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-200-227 External Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-227 Supplier Lumos Diagnostics Supplier No. CP0007

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FebriDx External Controls are used in the FebriDx test as assayed quality control samples to assess the performance and reliability of the FebriDx test.

  • FebriDx External Controls are used in the FebriDx test as assayed quality control samples to assess the performance and reliability of the FebriDx test.
  • Positive and negative external controls should be used as consistent with good laboratory practice.
  • The manufacturer recommends external controls be run with every new lot, every new shipment, and with every first time operator, but every testing site should follow their own protocols for running external controls.

Specifications

For Use With (Equipment) FebriDx test
Type External Control
Disposable Single-use
Level 2
Sterility Non-sterile
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.