Learn More
Medchemexpress LLC Febrifugine dihydrochloride | 32434-42-7 | 25 MG
SDP
Catalog No. 5000421933
Encompass_Preferred
Febrifugine dihydrochloride is a quinazolinone alkaloid that demonstrates antimalarial activity through its inhibition of *P. falciparum*. Beyond its antimalarial properties, this compound also exhibits an ability to inhibit bladder cancer by suppressing DNA synthesis, promoting apoptosis, and reducing steroid synthesis.
  • Inhibits *P. falciparum*.
  • Acts as an antimalarial agent.
  • Inhibits bladder cancer cell proliferation.
  • Induces apoptosis in bladder cancer cells.
  • Reduces steroid synthesis in bladder cancer cells.
Catalog No. 50-004-21933 Supplier Medchemexpress LLC Supplier No. 25MGHYN2384A
Febrifugine dihydrochloride is a quinazolinone alkaloid that demonstrates antimalarial activity through its inhibition of *P. falciparum*. Beyond its antimalarial properties, this compound also exhibits an ability to inhibit bladder cancer by suppressing DNA synthesis, promoting apoptosis, and reducing steroid synthesis.
  • Inhibits *P. falciparum*.
  • Acts as an antimalarial agent.
  • Inhibits bladder cancer cell proliferation.
  • Induces apoptosis in bladder cancer cells.
  • Reduces steroid synthesis in bladder cancer cells.