Learn More
Medchemexpress LLC Febrifugine dihydrochloride | 32434-42-7 | 50 MG
SDP
Catalog No. 5000421355
Encompass_Preferred
Febrifugine dihydrochloride is a quinazolinone alkaloid with antimalarial and anticancer activity. It inhibits Plasmodium falciparum and also inhibits bladder cancer by hindering DNA synthesis, inducing apoptosis, and reducing steroids.
  • Exhibits antimalarial activity against P. falciparum.
  • Inhibits proliferation of bladder cancer cells.
  • Suppresses DNA synthesis and induces cell death in cancer cells.
  • Reduces steroidogenesis and promotes apoptosis.
  • Significantly reduces mortality and parasitemia levels in infected mice.
  • Effectively inhibits bladder cancer xenograft proliferation in vivo.
Catalog No. 50-004-21355 Supplier Medchemexpress LLC Supplier No. HYN2384A50MG
Febrifugine dihydrochloride is a quinazolinone alkaloid with antimalarial and anticancer activity. It inhibits Plasmodium falciparum and also inhibits bladder cancer by hindering DNA synthesis, inducing apoptosis, and reducing steroids.
  • Exhibits antimalarial activity against P. falciparum.
  • Inhibits proliferation of bladder cancer cells.
  • Suppresses DNA synthesis and induces cell death in cancer cells.
  • Reduces steroidogenesis and promotes apoptosis.
  • Significantly reduces mortality and parasitemia levels in infected mice.
  • Effectively inhibits bladder cancer xenograft proliferation in vivo.