Learn More
Apexbio Technology LLC Febuxostat, 5mg. Cas: 144060-53-7 MFCD: MFCD00871598. non-purine selective inhibitor of xanthine oxidase
SDP
Catalog No. 501904963
Encompass_Preferred
IC50: Febuxostat displayed potent mixed-type inhibition of the activity of xanthine oxidase (XO), with Ki value of 0.6 nM [1]. Febuxostat was also reported to be 1000-fold (IC50=1.8 nM) more potent than allopurinol (IC50= 2.9 M) at inhibiting XO-dependent uric acid formation [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4963 Supplier Apexbio Technology LLC Supplier No. A59265
IC50: Febuxostat displayed potent mixed-type inhibition of the activity of xanthine oxidase (XO), with Ki value of 0.6 nM [1]. Febuxostat was also reported to be 1000-fold (IC50=1.8 nM) more potent than allopurinol (IC50= 2.9 M) at inhibiting XO-dependent uric acid formation [2]. Other sizes are available. Please inqury us for quote.