Learn More
Cayman Chemical Febuxostat Acyl GlucuronID
SDP
Catalog No. 502935124
Encompass_Preferred
A metabolite of febuxostat; is formed via glucuronidation of febuxostat by UGTs
Catalog No. 50-293-5124 Supplier Cayman Chemical Supplier No. 26599500 UG
A metabolite of febuxostat; is formed via glucuronidation of febuxostat by UGTs