Learn More
Medchemexpress LLC Febuxostat-d7 | 1285539-74-3 | 99.1% | C16H9D7N2O3S | 1 MG
SDP
Catalog No. 5000353713
Encompass_Preferred
Febuxostat-d7 is a deuterium labeled form of Febuxostat, a selective xanthine oxidase inhibitor with a Ki of 0.6 nM. It serves as a tracer and an internal standard for quantitative analysis. Deuterated compounds are used as tracers for quantitation during drug development and can affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled Febuxostat
  • Selective xanthine oxidase inhibitor with a Ki of 0.6 nM
  • For research use only
Catalog No. 50-003-53713 Supplier Medchemexpress LLC Supplier No. HY14268S11MG
Febuxostat-d7 is a deuterium labeled form of Febuxostat, a selective xanthine oxidase inhibitor with a Ki of 0.6 nM. It serves as a tracer and an internal standard for quantitative analysis. Deuterated compounds are used as tracers for quantitation during drug development and can affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled Febuxostat
  • Selective xanthine oxidase inhibitor with a Ki of 0.6 nM
  • For research use only